missing translation for 'onlineSavingsMsg'
Learn More

Irganox 1141 (>85%), TRC

Product Code. 30008517
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008517 10 mg 10mg
30007229 50 mg 50mg
30005220 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008517 Supplier Toronto Research Chemicals Supplier No. TRCI10121510MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134701-20-5
Molecular Formula C24H42O
Molecular Weight (g/mol) 346.59
Synonym 2-(Hexadecan-2-yl)-4,6-dimethylphenol;2,4-Dimethyl-6-(1-methylpentadecyl)phenol
IUPAC Name 2-hexadecan-2-yl-4,6-dimethylphenol
SMILES CCCCCCCCCCCCCCC(C)c1cc(C)cc(C)c1O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C24H42O/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-21(3)23-19-20(2)18-22(4)24(23)25/h18-19,21,25H,5-17H2,1-4H3
Formula Weight 346.3236
Chemical Name or Material Irganox 1141
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.