Learn More
IRAK inhibitor 4, MedChemExpress
Product Code. 16488307
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488307 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16488307 Supplier Medchem Express Supplier No. HY13278
 Restricted Item
Estimated Shipment: 28-09-2026
MedChemExpress IRAK inhibitor 4 is an interleukin-1 receptor associated kinase 4(IRAK4) inhibitor.
IRAK inhibitor 4 is an interleukin-1 receptor associated kinase 4(IRAK4) inhibitor.
Specifications
Chemical Name or Material IRAK inhibitor 4
CAS 1012104-68-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C33H35F3N6O3
Quantity 5 mg
Solubility Information DMSO : 12.5 mg/mL (20.14 mM; Need ultrasonic)
SMILES OC1CCC(N2C3=C(C=C(C=C3)C(NCCC(C)C)=O)N=C2NC4=NNC5=C4C=C(C=C5)C6=CC=CC=C6OC(F)(F)F)CC1
Molecular Weight (g/mol) 620.66
Formula Weight 620.66
Percent Purity 98.05%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation