Learn More
IRAK inhibitor 3, MedChemExpress
Product Code. 16478307
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478307 5 mg 5mg
16425618 10 mg 10mg
16411787 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16478307 Supplier Medchem Express Supplier No. HY13277
 Restricted Item
Estimated Shipment: 02-11-2026
MedChemExpress IRAK inhibitor 3 is an interleukin-1 (IL-I) receptor-associated kinase (IRAK) kinase modulator extracted from patent WO2008030579 A2.
IRAK inhibitor 3 is an interleukin-1 (IL-I) receptor-associated kinase (IRAK) kinase modulator extracted from patent WO2008030579 A2.
Specifications
Chemical Name or Material IRAK inhibitor 3
CAS 1012343-93-9
Color Gray
Purity Grade Notes Research
Molecular Formula C21H21N5O4S
Quantity 5 mg
Solubility Information DMSO : 13 mg/mL (29.58 mM; Need ultrasonic and warming)
SMILES O=S(C1=CC=C(CNC2=NN3C(C4=CC(OC)=C(C=C4)OC)=CN=C3C=C2)C=C1)(N)=O
Molecular Weight (g/mol) 439.49
Formula Weight 439.49
Percent Purity 98.17%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation