Learn More
Thermo Scientific IQOQPQ Package for ULT and CRYO -150°C to -50°C
Product Code. 32176307 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Thermo Scientific ULT Freezers
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
32176307 Thermo Scientific ULT Freezers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32176307 Supplier Thermo Scientific Supplier No. IQOQPCKULTMVS
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Læg i indkøbskurv
Denne vare kan ikke returneres. Se returpolicy
Provides qualification execution/support for compressor-based ULT and cryogenic freezers, including units where LN2 backup is secondary to the primary cooling system.
  • Not suitable for liquid nitrogen Dewar-type cryopreservation systems
  • Confirm whether documents are included/supplied free of charge or require separate document-pack logic
TRUSTED_SUSTAINABILITY
Tekniske data
For Use With (Equipment) Thermo Scientific ULT Freezers
Product Type IQ/OQ/PQ with Completion of Documents
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.