Learn More
Memmert™ Iq/Oq Document With Device-Specific Works Test Data (1 Temperature Value), For Un160Pa
Shop All Memmert ProductsDescription
- IQ/OQ document with works test data for chamber
- 9 measuring points (size 30), 27 measuring points (sizes 55 - 1060) to DIN 12880:2007-05 (further temperature values on demand), PQ check list as support for validation by customer
Specifications
Specifications
| Product Type | IQ/OQ Documentation |