Learn More
Memmert™ Iq/Oq Document W/Device-Specific Works Test Data (Temperature And Vacuum Value), For Vo49
Shop All Memmert ProductsDescription
- Temperature distribution survey for 5 measuring points to DIN 12880:2007-05 as support for validation by customer (VO, VOcool only) valid for one thermo shelf