missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ IQ/OQ Document with Test Data for UN75plus

Product Code. 17590526 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
UN75plus Oven
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17590526 UN75plus Oven Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17590526 Supplier Memmert™ Supplier No. UN75plusD00127
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

IQ/OQ document with temperature distribution survey for one free-selectable temperature value.

  • Includes temperature distribution survey with 9 temperature measuring points (size 30) and 27 measuring points (sizes 55 - 1060) to DIN 12880:2007-05 (further temperature values on demand)
  • PQ check list as support for validation by customer

Specifications

For Use With (Equipment) UN75plus Oven
Product Type IQ/OQ Documentation
Includes Test Data
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.