Learn More
Memmert™ IQ/OQ Document with Test Data for UF260plus
Product Code. 17509175 Shop All Memmert Products
Click to view available options
:
For Use With: UF260plus
Product Code. 17509175
Brand: Memmert™ UF260plusD00127
IQ/OQ document with temperature distribution survey for one free-selectable temperature value.
  • Includes temperature distribution survey with 9 temperature measuring points (size 30) and 27 measuring points (sizes 55 - 1060) to DIN 12880:2007-05 (further temperature values on demand)
  • PQ check list as support for validation by customer
Specifications
For Use With (Equipment) UF260plus Oven
Product Type IQ/OQ Documentation
Includes Test Data