Learn More
Memmert™ IQ/OQ Document with Test Data for IF55
Product Code. 17547085 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
IF55 Incubator
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17547085 IF55 Incubator Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17547085 Supplier Memmert™ Supplier No. IF55D00127
 Restricted Item
Estimated Shipment: 14-12-2026
IQ/OQ document with temperature distribution survey for one free-selectable temperature value.
  • Includes temperature distribution survey with 9 temperature measuring points (size 30) and 27 measuring points (sizes 55 - 1060) to DIN 12880:2007-05 (further temperature values on demand)
  • PQ check list as support for validation by customer
Specifications
For Use With (Equipment) IF55 Incubator
Product Type IQ/OQ Documentation
Includes Test Data