Learn More
Memmert™ IQ/OQ Document with Device-Specific Works Test Data- Temperature and Humidity
Product Code. 17119824 Shop All Memmert Products
Click to view available options
Unit Size:
Each
For Use With (Equipment):
HPP110eco
HPP260eco
HPP750eco
Product Code. 17119824
Brand: Memmert™ HPP410ecoD00137
For one free-selectable temperature and humidity value, and measuring of light intensity
  • Includes temperature distribution survey at Memmert
  • For 27 measuring points to DIN 12880:2007-05
  • PQ check list as support for validation by customer.
Specifications
Product Type IQ/OQ Document with Device-specific Works Test Data