Learn More
Memmert™ IQ/OQ Document with Device-Specific Works Test Data - Temperature
Product Code. 17190215 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP1400eco
HPP2200eco
HPP410eco
HPP750eco
IPP1060eco
IPP1400eco
IPP2200eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17190215 IPP410eco Each
17100135 IPP1400eco Each
17100185 IPP260eco Each
17109684 HPP110eco Each
17110155 IPP2200eco Each
17110265 IPP750eco Each
17129744 HPP2200eco Each
17149774 - Each
17170055 IPP1060eco Each
17179864 HPP750eco Each
17189714 HPP1400eco Each
17199634 HPP1060eco Each
17199814 HPP410eco Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 17190215 Supplier Memmert™ Supplier No. IPP410ecoD00127
 Restricted Item
Estimated Shipment: 19-10-2026
For one free-selectable temperature value, Includestemperature distribution survey at Memmert to DIN 12880:2007-05
  • 27 measuring points (sizes 110 - 1060, 2200)
  • 26 measuring points (size 1400)
  • PQ checklist as support for validation by customer
Specifications
For Use With (Equipment) IPP410eco
Product Type IQ/OQ Document with Device-specific Works Test Data