Learn More
Memmert™ IQ/OQ Document with Device-Specific Works Test Data - CO2, Humidity and Temperature
Product Code. 17169924 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
ICO105
ICO150
ICO240
ICO50
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17169924 ICO105 Each
17139994 ICO240 Each
17160025 ICO50 Each
17199954 ICO150 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17169924 Supplier Memmert™ Supplier No. ICO105D38897
 Restricted Item
Estimated Shipment: 14-12-2026
For one free-selectable CO2, humidity and temperature value
  • Includes temperature distribution survey at Memmert
  • For 27 measuring points to DIN 12880:2007-05
  • PQ check list as support for validation by customer.
Specifications
For Use With (Equipment) ICO105
Product Type IQ/OQ Document with Device-specific Works Test Data