Description
- Includes temperature distribution survey at Memmert
- For 9 measuring points to DIN 12876.2
Specifications
Specifications
| For Use With (Equipment) | WTB50 |
| Product Type | IQ/OQ Document with Device-specific Works Test Data |