Learn More
Memmert™ Iq Document With Device-Specific Works Test Data For Validation By Customer, For Ipp30
Product Code. 16367457 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
30, 55, 75, 110, 160, 260, 450, 750, 1060 Models Climate Chamber
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
16367457 30, 55, 75, 110, 160, 260, 450, 750, 1060 Models Climate Chamber Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16367457 Supplier Memmert™ Supplier No. IPP30D00124
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Includes
Works test data
Includes IQ document with works test data for chamber.
  • OQ/PQ check list as support for validation by customer
Specifications
For Use With (Equipment) 30, 55, 75, 110, 160, 260, 450, 750, 1060 Models Climate Chamber
Product Type IQ Document