Learn More
Memmert™ IQ Document with Test Data for UN160
Product Code. 17549865 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
UN160 Oven
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17549865 UN160 Oven Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17549865 Supplier Memmert™ Supplier No. UN160D00124
 Restricted Item
Estimated Shipment: 26-10-2026
IQ document with device-specific works test data for validation by customer.
  • OQ/PQ check list as support for validation by customer
Specifications
For Use With (Equipment) UN160 Oven
Product Type IQ Document
Includes Test Data