Learn More
Memmert™ IQ Document with Test Data for SN260plus
Product Code. 17528565 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
SN260plus Sterilizer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17528565 SN260plus Sterilizer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17528565 Supplier Memmert™ Supplier No. SN260plusD00124
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
IQ document with device-specific works test data for validation by customer.
  • OQ/PQ check list as support for validation by customer
Specifications
For Use With (Equipment) SN260plus Sterilizer
Product Type IQ Document
Includes Test Data