Learn More
Memmert™ IQ Document with Test Data for IF260plus
Product Code. 17516845 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
IF260plus Incubator
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17516845 IF260plus Incubator Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17516845 Supplier Memmert™ Supplier No. IF260plusD00124
IQ document with device-specific works test data for validation by customer.
  • OQ/PQ check list as support for validation by customer
Specifications
For Use With (Equipment) IF260plus Incubator
Product Type IQ Document
Includes Test Data