Learn More
Ipronidazole-d3 (1-methyl-d3), CDN
Product Code. 30318453
Change view
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30318453 0.005 g 0.01g
30311644 0.01 g 0.01g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30318453 Supplier CDN Supplier No. CDND81880.005G
 Restricted Item
Estimated Shipment: 10-11-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ipronidazole D3
CAS 1015855-83-0
Molecular Formula C7 2H3 H8 N3 O2
Quantity 0.005 g
Synonym 1 H-Imidazole, 1-(methyl-d3)-2-(1-methylethyl)-5-nitro-, Ipronidazole D3, Ipronidazole D3 (N-methyl D3)
InChI Formula InChI=1 S/C7H11N3O2/c1-5(2)7-8-4-6(9(7)3)10(11)12/h4-5 H,1-3H3/i3D3
SMILES [2 H]C([2 H])([2 H])n1c(ncc1[N+](=O)[O-])C(C)C
IUPAC Name 5-nitro-2-propan-2-yl-1-(trideuteriomethyl)imidazole
Molecular Weight (g/mol) 172.2
Formula Weight 172.104 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity