missing translation for 'onlineSavingsMsg'
Learn More

Iopromid 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30319229
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30319229 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30319229 Supplier Dr. Ehrenstorfer Supplier No. DREA14348600AL100
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Iopromide
CAS 73334-07-3
Analyte or Component Names Iopromide
Shipping Condition Room Temperature
Molecular Formula C18 H24 I3 N3 O8
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Iopromide, N,N'-Bis(2,3-dihydroxypropyl)-2,4,6-triiodo-5-[(methoxyacetyl)amino]-N-methylbenzene-1,3-dicarboxamide, 1,3-Benzenedicarboxamide, N1,N3-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-5-[(2-methoxyacetyl)amino]-N1-methyl-, 1,3-Benzenedicarboxamide, N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-5-[(methoxyacetyl)amino]-N-methyl- (9CI), Iopromide, Iopromidum, SHL 414C, Ultravist, Ultravist 300, Ultravist 370, ZK 35760
InChI Formula InChI=1S/C18H24I3N3O8/c1-24(4-9(28)6-26)18(31)12-13(19)11(17(30)22-3-8(27)5-25)14(20)16(15(12)21)23-10(29)7-32-2/h8-9,25-28H,3-7H2,1-2H3,(H,22,30)(H,23,29)
SMILES COCC(=O)Nc1c(I)c(C(=O)NCC(O)CO)c(I)c(C(=O)N(C)CC(O)CO)c1I
IUPAC Name 1-N,3-N-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-5-[(2-methoxyacetyl)amino]-3-N-methylbenzene-1,3-dicarboxamide
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 791.11
Quantity 1 mL
Formula Weight 790.8697 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.