Learn More
Sartorius™ Ionizing Blower
Product Code. 10459312 Shop All Sartorius Products
Change view
Click to view available options
230V, 50Hz
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10459312 230V, 50Hz
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10459312 Supplier Sartorius™ Supplier No. YIB010DR
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Neutralize static electricity within seconds. Sartorius™ Ionizing Blower works by shooting ions at the material to be weighed.
  • Neutralizes static electricity
  • Eliminates the need for complicated, expensive, and often ineffective humidity change approaches.
TRUSTED_SUSTAINABILITY
Specifications
Electrical Requirements 230 V, 50 Hz
Weight (Metric) 0.6 kg