missing translation for 'onlineSavingsMsg'
Learn More

Iodoethane-d5 (stabilized with Copper wire), TRC

Product Code. 30406374
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30406374 5 g -
30397081 1 g 1g
30405200 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30406374 Supplier Toronto Research Chemicals Supplier No. TRCI7058725G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6485-58-1
Molecular Formula C2 2H5 I
Molecular Weight (g/mol) 161
Synonym Ethane-1,1,1,2,2-d5, 2-iodo- (ACI), 2-Iodoethane-1,1,1,2,2-d5 (ACI), Ethane-d5, iodo- (7CI, 8CI, 9CI), 1,1,1,2,2-Pentadeuterio-2-iodoethane, Ethyl-d5 iodide, Iodoethane-d5, Iodoethane-d5, Iodopentadeuterioethane, Pentadeuterioethyl iodide, Pentadeuteroethyl iodide, Iodoethane-D5
IUPAC Name 1,1,1,2,2-pentadeuterio-2-iodoethane
SMILES [2H]C([2H])([2H])C([2H])([2H])I

Specifications

Quantity 5 g
InChI Formula InChI=1S/C2H5I/c1-2-3/h2H2,1H3/i1D3,2D2
Formula Weight 160.975
Percent Purity >95%
Chemical Name or Material Iodoethane-d5
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.