missing translation for 'onlineSavingsMsg'
Learn More

Iodine Monochloride, Sigma-Aldrich™

Product Code. 15834161
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15834161 250 g 250g
15824161 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15834161 Supplier Sigma Aldrich Supplier No. 8047710250
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-99-0

Specifications

Boiling Point 97°C/1013 hPa (decomposition)
Melting Point 14°C to 27°C
Quantity 250 g
MDL Number MFCD00011354
Synonym Iodine monochloride, Iodine chloride, Chlorine iodide, Iodine monochloride
Molecular Weight (g/mol) 162.36
Grade Research
Density 3.1 g/cm3 at 29°C
Chemical Name or Material Iodine Monochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.