Learn More
Iodide ISE Standard Solution, Mettler Toledo™
Product Code. 11977521
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11977521 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11977521 Supplier Mettler Toledo Online Supplier No. 51344776
  • ONLINE EXCLUSIVE PRICE
    133.00 EUR
    131.01 EUR / 500mL
    Save 1.99 EUR
    1% Off
 Restricted Item
In Stock
Specifications
Description ISE standard I 1000 mg/L, 500mL
Packaging 500 mL
Physical Form Liquid
Color Colorless
Quantity 500 mL