Learn More
Invitrogen™ pBudCE4.1 Mammalian Expression Vector
Description
Beinhaltet
20μg each of pBudCE4.1 and pBudCE4.1/lacZ/CAT positive control vectors are supplied supercoiled and lyophilized.
- Using pBudCE4.1 to generate stable mammalian expression cell lines ensures that there is an equivalent copy number of each gene in cell; this can eliminate variable expression due to differences in gene copy number
- Provides expression cassettes with the CMV promoter for high-level transcription of genes with optional c-myc epitope tag for rapid detection and 6xHis sequence for simple purification
- Human EF-1α promoter for high-level expression of genes with optional V5 epitope tag for rapid detection and 6xHis sequence for simple purification
- Sh ble (ZeoR) gene for efficient selection in both mammalian cells and E. coli with selection agent Zeocin™
Constitutive Expression, Mammalian Expression, Multi-Gene Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
Spezifikation
Spezifikation
| Constitutive or Inducible System | Constitutive |
| Delivery Type | Transfection |
| Promoter | EF-1α, CMV |
| Product Type | Mammalian Expression Vector |
| Selection Agent (Eukaryotic) | Zeocin™ |
| Content And Storage | 20 μg each of pBudCE4.1 and pBudCE4.1/lacZ/CAT positive control vectors are supplied supercoiled and lyophilized. Store at -20°C. Vectors are guaranteed stable for 6 months when properly stored. |
| Protein Tag | His Tag (6x), V5 Epitope Tag, c-Myc Epitope Tag |
| Cloning Method | Restriction Enzyme/MCS |
| Quantity | 20 μg |
| Vector | pBud |
| Mehr anzeigen |