Learn More
Invitrogen™ LR Clonase II for BaculoDirect™ Kits
Click to view available options
Quantity:
10 reactions
Unit Size:
10 reactions
Description
- Combination of Gateway Technology with baculovirus expression makes BaculoDirect System fastest and easiest method for production of recombinant baculovirus
Cloning, Gateway Cloning, Gateway Recombination
Order Info
Shipping Conditions: Dry ice
Specifications
Specifications
| Content And Storage | Store LR Clonase™ mix at -80°C. All components are guaranteed stable for 6 months if stored properly. |
| Shipping Condition | Dry Ice |
| Enzyme | LR Clonase |
| Quantity | 10 reactions |
| Product Type | LR Clonase II |
| Product Line | BaculoDirect, Clonase, Gateway |