missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 2b beta 3, Rabbit anti-Human, Clone: 1000000, Novus Biologicals™
Rabbit Monoclonal Antibody
275.00 EUR - 628.00 EUR
Specifications
| Antigen | Integrin alpha 2b beta 3 |
|---|---|
| Clone | 1000000 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, ELISA, Immunocytochemistry/Immunofluorescence |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18285809
|
Novus Biologicals
NBP2-81080-0.025MG |
0.025 mg |
275.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18227939
|
Novus Biologicals
NBP2-81080-0.2MG |
0.2 mg |
628.00 EUR
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Integrin alpha 2b beta 3 Monoclonal specifically detects Integrin alpha 2b beta 3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Integrin alpha 2b beta 3 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| 3674 | |
| Human blood platelets. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 1000000 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| ITGA2B | |
| IgG κ | |
| Protein A purified | |
| The antibody binds to Integrin alpha 2b beta 3 with an affinity (Kd) of 16.2+-6 nM. The antibody completely inhibits platelet aggregation at 7.8 ug/ml. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title