missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 2b beta 3, Rabbit anti-Human, Clone: 1000000, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81080-0.025MG
This item is not returnable.
View return policy
Description
Integrin alpha 2b beta 3 Monoclonal specifically detects Integrin alpha 2b beta 3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Integrin alpha 2b beta 3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| ITGA2B | |
| Human blood platelets. | |
| 0.025 mg | |
| Primary | |
| The antibody binds to Integrin alpha 2b beta 3 with an affinity (Kd) of 16.2+-6 nM. The antibody completely inhibits platelet aggregation at 7.8 ug/ml. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG κ |
| Western Blot, ELISA, Immunofluorescence | |
| 1000000 | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 3674 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction