Learn More
Fisherbrand™ Insert For Maxi Boxes
Product Code. 12367167 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Tube Size:
8 x 1.5mL
15 x 5mL
8 x 20mL
8 x 50mL
15 x 15mL
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tube Size Unit Size
12367167 8 x 50mL Each
10315311 15 x 15mL Each
12327167 8 x 20mL Each
12347167 8 x 1.5mL Each
12357167 15 x 5mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12367167
Estimated Shipment: 09-04-2027
Add to basket
Add to basket
Manufactured with hinged lids and accommodate interchangeable inserts that hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes; these serve as an ideal solution for short-term storage of radioactive waste or radioisotopes.
  • Inserts for Maxi Boxes
  • Cleaver Scientific storage boxes are manufactured with hinged lids and accommodate interchangeable inserts that hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
  • Also supplied is our range of floor-standing and benchtop bins with anti-slip feet and hinged lids
  • These serve as an ideal solution for short-term storage of radioactive waste or radioisotopes
  • Both the Beta and Gamma storage bins are available in five sizes, while the two largest bin models have wheels for easy transportation
  • Optional heavy duty drawstring bags may also be purchased
Specifications
For Use With (Equipment) For Maxi Boxes
Height (Metric) 40mm
Product Type Insert
Tube Size 8 x 50mL
Width (Metric) 160mm