Learn More
Miele Professional™ Insert
Product Code. 13192270 Shop All Miele Products
Change view
Click to view available options
Height (Metric):
95 mm
120 mm
145 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
13192270 120 mm Each
13112280 95 mm Each
13102280 145 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 13192270 Supplier Miele Professional™ Supplier No. 3830270
  • ONLINE EXCLUSIVE PRICE
    537.00 EUR
    415.00 EUR / Pack of 1
    Save 122.00 EUR
    22% Off
Estimated Shipment: 02-11-2026
Specifications
Product Type Insert
For Use With (Equipment) 38 Petri dishes with diameter 100mm, for use in upper or lower basket
Depth (Metric) 445 mm
Height (Metric) 120 mm
Width (Metric) 460 mm
Quantity 1 Each