Learn More
Miele Professional™ Insert
Product Code. 13112280 Shop All Miele Products
Change view
Click to view available options
Height (Metric):
95 mm
120 mm
145 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) unitSize
13112280 95 mm Each
13192270 120 mm Each
13102280 145 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 13112280 Supplier Miele Professional™ Supplier No. 3830290
Estimated Shipment: 01-10-2026
Specifications
Product Type Insert
For Use With (Equipment) G 7883, G 7893, G 7883 CD, G 7835 CD, G 7836 CD, 56 holders, 70mm H, spacing ca. 26mm
Depth (Metric) 445 mm
Height (Metric) 95 mm
Width (Metric) 485 mm
Quantity 1 Each