missing translation for 'onlineSavingsMsg'
Learn More

inS3-54A18, MedChemExpress

Product Code. 16470177
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16470177 50 mg 50mg
16490076 2 mg 2mg
16473798 10 mg 10mg
16426527 5 mg 5mg
16424856 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16470177 Supplier Medchem Express Supplier No. HY103128
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress inS3-54A18 is a potent STAT3 inhibitor, with anti-cancer properties.

inS3-54A18 is a potent STAT3 inhibitor, with anti-cancer properties.

Specifications

Chemical Name or Material inS3-54A18
CAS 328998-53-4
Color Orange
Purity Grade Notes Research
Molecular Formula C23H16ClNO2
Quantity 50 mg
Solubility Information DMSO : ≥ 150 mg/mL (401.25 mM)
SMILES O=C1N(C2=CC=C(O)C=C2)C(C3=CC=CC=C3)=C/C1=C/C4=CC=C(Cl)C=C4
Molecular Weight (g/mol) 373.83
Formula Weight 373.83
Percent Purity 99.83%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.