missing translation for 'onlineSavingsMsg'
Learn More

Inpyrfluxam, TRC

Product Code. 30138298
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138298 250 mg 250mg
30136933 500 mg 500mg
17903285 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138298 Supplier Toronto Research Chemicals Supplier No. TRCI854065250MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1352994-67-2
Molecular Formula C18 H21 F2 N3 O
Molecular Weight (g/mol) 333.38
Synonym 3-(Difluoromethyl)-N-[(3R)-2,3-dihydro-1,1,3-trimethyl-1H-inden-4-yl]-1-methyl-1H-pyrazole-4-carboxamide, 3-(Difluoromethyl)-1-methyl-N-((3R)-1,1,3-trimethyl-2,3-dihydro-1H-inden-4-yl)-1H-pyrazole-4-carboxamide, 3-(Difluoromethyl)-1-methyl-N-[(3R)-1,1,3-trimethyl-2,3-dihydro-1H-inden-4-yl]-1H-pyrazole-4-carboxamide, Inpyrfluxam
IUPAC Name 3-(difluoromethyl)-1-methyl-N-[(3R)-1,1,3-trimethyl-2,3-dihydroinden-4-yl]pyrazole-4-carboxamide
SMILES C[C@@H]1CC(C)(C)c2cccc(NC(=O)c3cn(C)nc3C(F)F)c12

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C18H21F2N3O/c1-10-8-18(2,3)12-6-5-7-13(14(10)12)21-17(24)11-9-23(4)22-15(11)16(19)20/h5-7,9-10,16H,8H2,1-4H3,(H,21,24)/t10-/m1/s1
Formula Weight 333.1653
Percent Purity >95%
Chemical Name or Material Inpyrfluxam
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.