Learn More
WTW™ inoLab™ 7110 Benchtop pH Meter
Click to view available options
Probe Type:
3 in 1 pH electrode
Combined pH electrode
Not included
Unit Size:
Each
Description
- Repeatable measuring results by automatic recognition of stable values
- Secure operation: automated functions reduce the number of keys
- Increased measuring accuracy by calibration timer with periodic reminders for calibration
- Highly suitable for routine measurements in the lab, where automatic documentation is not a priority
- With smooth, easy to clean surface
- Available as a single meter, as a set with standard electrode with built-in T sensor, or as a set with a precision electrode
Specifications
Specifications
| Includes | DIN socket, Universal power supply, stand and operation manual, Combined pH electrode SenTix™ 42/41, buffer 4, 7 and 10.01, 3mol/L KCl, WTW technical buffers, DIN/NIST buffers |
| Accuracy (Temperature) | ±0.1K |
| Accuracy (mV) | ±0.3, ±1.0 |
| Accuracy (pH) | ±0.005, ±0.01 |
| Data Management Details | Current record calibration memory |
| For Use With (Application) | Ideal for routine measurements in the laboratory where automatic documentation is not the priority and for AC and battery operation |
| Range (pH) | -2 to 20 |
| Resolution (pH) | -2 to 20 ±0.01, -2 to 19.999 ±0.005 |
| Range (mV) | ±1200, ±0.3 |
| Resolution (mV) | ±(1200.0 ±0.3mV) |
| Show More |