Learn More
Greiner Bio-One Disposable Inoculation Loops and Needles
Product Code. 10217591
Change view
Click to view available options
Volume:
1 μL
Product Type:
Inoculating Loop
Color:
Blue
White
Packaging Type:
Bulk
Individually Wrapped
Unit Size:
Pack of 2000
Pack of 600
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume Product Type Color Packaging Type Unit Size
10217591 1 μL Inoculating Loop White Individually Wrapped Pack of 600
10792142 - Inoculating Loop Blue Bulk Pack of 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10217591 Supplier Greiner Bio-One Supplier No. 731161
  • ONLINE EXCLUSIVE PRICE
    169.00 EUR
    131.86 EUR / Pack of 600
    Save 37.14 EUR
    21% Off
Estimated Shipment: 11-01-2027
Manufactured from polystyrene for strength and flexibility. Greiner Bio-One™ Disposable Inoculation Loops provide easier collection and inocluation,
  • Needles are ideal for picking up single colonies; Loops are ideal for picking up designated volume of culture
  • Packaged individually or bulk
  • Hydrophobic
  • Temperature range: –20 to +60°C
  • Sterilized by irradiation
  • For single use only
TRUSTED_SUSTAINABILITY
Specifications
Disposable Single-use
Volume 1 μL
Length (English) 7.87 in.
Length (Metric) 200 mm
Material Polystyrene
Sterility Sterile
Size Standard
Color White
Product Type Inoculating Loop
Packaging Type Individually Wrapped