Learn More
KARTELL Industrial Tank
Product Code. 10501515
Change view
Click to view available options
Capacity (Metric):
20 L
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10501515 20 L Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10501515 Supplier KARTELL Supplier No. 0178300
  • ONLINE EXCLUSIVE PRICE
    164.00 EUR
    126.60 EUR / Pack of 6
    Save 37.40 EUR
    22% Off
View all promotions
 Restricted Item
Estimated Shipment: 21-12-2026
Industrial Tanks are specifically designed for stacking to promote stability during transportation for industry and laboratory practice.
  • Standardized industrial packaging design, and even if specifically addressed to a use in industrial packaging, they could result useful in everyday laboratory practice.
  • They are made of High Density PE food grade, with a comfortable handle, and feature a secure seal-cap following DIN recommendations.
  • They are conform to UN regulation on transportation of dangerous and toxic liquids and are suitable for road (ADR-RID), sea (IMO-IMDG) and air (IATA-ICAO) transport.
  • SUITABLE FOR FOODSTUFF.
Specifications
Closure Included
Color White
Material Polyethylene
Capacity (Metric) 20 L
No. per Pack 6 pcs.