Learn More
3M™ Maintenance Sorbent Pads M-A2002
Product Code. 11767184
Change view
Click to view available options
Length (Metric):
520 mm
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
11767184 520 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11767184 Supplier 3M™ Supplier No. 7100003876
3M™ Maintenance Sorbent Pads are designed to be used in and around the work area to catch leaks and drips, pads are highly absorbent and simple to use.
  • Sorbent pads designed for use in and around the work area to catch leaks and drips
  • For use on most non-aggressive liquids
  • Very easy to use and to retrieve
  • Promotes a cleaner, safer working environment
Order Info
dimProductType
TRUSTED_SUSTAINABILITY
Specifications
Color Grey
Type Sorbent Pads
Length (Metric) 520 mm
Width (Metric) 400 mm