missing translation for 'onlineSavingsMsg'
Learn More

Indolicidin TFA Salt, TRC

Product Code. 30405320
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30405320 1 mg 1mg
30402706 5 mg 5mg
30400267 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30405320 Supplier Toronto Research Chemicals Supplier No. TRCI6272731MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Indolicidin TFA Salt
Molecular Formula C100H132N26O13 • x(C2HF3O2)
Quantity 1 mg
Synonym L-Isoleucyl-L-leucyl-L-prolyl-L-tryptophyl-L-lysyl-L-tryptophyl-L-prolyl-L-tryptophyl-L-tryptophyl-L-prolyl-L-tryptophyl-L-arginyl-L-argininamide, 13: PN: WO03048383 SEQID: 2 claimed protein, 1: PN: WO2010043039 SEQID: 1 claimed protein, 1: PN: WO2010141135 SEQID: 6 claimed protein, 21: PN: WO0236612 SEQID: 28 unclaimed sequence, 26: PN: WO2007058336 SEQID: 26 claimed protein, 42: PN: US7381704 SEQID: 42 claimed sequence, 7: PN: WO2010149795 SEQID: 7 claimed protein
InChI Formula InChI=1S/C100H132N26O13.C2HF3O2/c1-5-57(4)85(102)95(136)123-79(45-56(2)3)96(137)124-42-20-36-82(124)92(133)118-76(46-58-51-110-68-28-11-6-23-63(58)68)89(130)116-74(33-16-17-39-101)88(129)121-80(49-61-54-113-71-31-14-9-26-66(61)71)97(138)125-43-21-38-84(125)94(135)120-78(48-60-53-112-70-30-13-8-25-65(60)70)91(132)122-81(50-62-55-114-72-32-15-10-27-67(62)72)98(139)126-44-22-37-83(126)93(134)119-77(47-59-52-111-69-29-12-7-24-64(59)69)90(131)117-75(35-19-41-109-100(106)107)87(128)115-73(86(103)127)34-18-40-108-99(104)105;3-2(4,5)1(6)7/h6-15,23-32,51-57,73-85,110-114H,5,16-22,33-50,101-102H2,1-4H3,(H2,103,127)(H,115,128)(H,116,130)(H,117,131)(H,118,133)(H,119,134)(H,120,135)(H,121,129)(H,122,132)(H,123,136)(H4,104,105,108)(H4,106,107,109);(H,6,7)/t57-,73-,74-,75-,76-,77-,78-,79-,80-,81-,82-,83-,84-,85-;/m0./s1
SMILES N=C(N)NCCC[C@@H](C(N)=O)NC([C@H](CCCNC(N)=N)NC([C@H](CC1=CNC2=CC=CC=C12)NC([C@H]3N(C([C@H](CC4=CNC5=CC=CC=C45)NC([C@H](CC6=CNC7=CC=CC=C67)NC([C@H]8N(C([C@H](CC9=CNC%10=CC=CC=C9%10)NC([C@H](CCCCN)NC([C@H](CC%11=CNC%12=CC=CC=C%11%12)NC([C@H]%13N(C([C@H](CC(C)C)NC([C@H]([C@@H](C)CC)N)=O)=O)CCC%13)=O)=O)=O)=O)CCC8)=O)=O)=O)CCC3)=O)=O)=O.O=C(O)C(F)(F)F
Molecular Weight (g/mol) 1906.2811402
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.