missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indolicidin TFA Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Indolicidin TFA Salt |
| Molecular Formula | C100H132N26O13 • x(C2HF3O2) |
| Quantity | 5 mg |
| Synonym | L-Isoleucyl-L-leucyl-L-prolyl-L-tryptophyl-L-lysyl-L-tryptophyl-L-prolyl-L-tryptophyl-L-tryptophyl-L-prolyl-L-tryptophyl-L-arginyl-L-argininamide, 13: PN: WO03048383 SEQID: 2 claimed protein, 1: PN: WO2010043039 SEQID: 1 claimed protein, 1: PN: WO2010141135 SEQID: 6 claimed protein, 21: PN: WO0236612 SEQID: 28 unclaimed sequence, 26: PN: WO2007058336 SEQID: 26 claimed protein, 42: PN: US7381704 SEQID: 42 claimed sequence, 7: PN: WO2010149795 SEQID: 7 claimed protein |
| InChI Formula | InChI=1S/C100H132N26O13.C2HF3O2/c1-5-57(4)85(102)95(136)123-79(45-56(2)3)96(137)124-42-20-36-82(124)92(133)118-76(46-58-51-110-68-28-11-6-23-63(58)68)89(130)116-74(33-16-17-39-101)88(129)121-80(49-61-54-113-71-31-14-9-26-66(61)71)97(138)125-43-21-38-84(125)94(135)120-78(48-60-53-112-70-30-13-8-25-65(60)70)91(132)122-81(50-62-55-114-72-32-15-10-27-67(62)72)98(139)126-44-22-37-83(126)93(134)119-77(47-59-52-111-69-29-12-7-24-64(59)69)90(131)117-75(35-19-41-109-100(106)107)87(128)115-73(86(103)127)34-18-40-108-99(104)105;3-2(4,5)1(6)7/h6-15,23-32,51-57,73-85,110-114H,5,16-22,33-50,101-102H2,1-4H3,(H2,103,127)(H,115,128)(H,116,130)(H,117,131)(H,118,133)(H,119,134)(H,120,135)(H,121,129)(H,122,132)(H,123,136)(H4,104,105,108)(H4,106,107,109);(H,6,7)/t57-,73-,74-,75-,76-,77-,78-,79-,80-,81-,82-,83-,84-,85-;/m0./s1 |
| SMILES | N=C(N)NCCC[C@@H](C(N)=O)NC([C@H](CCCNC(N)=N)NC([C@H](CC1=CNC2=CC=CC=C12)NC([C@H]3N(C([C@H](CC4=CNC5=CC=CC=C45)NC([C@H](CC6=CNC7=CC=CC=C67)NC([C@H]8N(C([C@H](CC9=CNC%10=CC=CC=C9%10)NC([C@H](CCCCN)NC([C@H](CC%11=CNC%12=CC=CC=C%11%12)NC([C@H]%13N(C([C@H](CC(C)C)NC([C@H]([C@@H](C)CC)N)=O)=O)CCC%13)=O)=O)=O)=O)CCC8)=O)=O)=O)CCC3)=O)=O)=O.O=C(O)C(F)(F)F |
| Molecular Weight (g/mol) | 1906.2811402 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?