Learn More
Indoleacetic Acid-13C6, TRC
Specifications
Specifications
| Chemical Name or Material | 1H-Indole-3-acetic Acid 13C6 (indole-3a,4,5,6,7,7a-13C6) |
| CAS | 100849-36-3 |
| Molecular Formula | 13C6 C4 H9 N O2 |
| Quantity | 10 mg |
| Synonym | 1H-Indole-3a,4,5,6,7,7a-13C6-3-acetic acid, 1H-Indole-3-acetic acid 13C6 (indole-3a,4,5,6,7,7a-13C6), 2-([3a,4,5,6,7,7a-13C6]-1H-indol-3-yl)acetic acid, 2-(3-Indolyl-3a,4,5,6,7,7a-13C6)acetic acid, 3-Indolylacetic acid-13C6 |
| InChI Formula | InChI=1S/C10H9NO2/c12-10(13)5-7-6-11-9-4-2-1-3-8(7)9/h1-4,6,11H,5H2,(H,12,13)/i1+1,2+1,3+1,4+1,8+1,9+1 |
| SMILES | OC(=O)Cc1c[nH][13c]2[13cH][13cH][13cH][13cH][13c]12 |
| IUPAC Name | 2-(1H-indol-3-yl)acetic acid |
| Molecular Weight (g/mol) | 181.14 |
| Formula Weight | 181.083 |
Product Title
Individuate un'opportunità di miglioramento?