missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-propionic acid, ∽99%, MP Biomedicals™

Product Code. 11358879 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Unit Size:
10g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358879 1 g 1g
11368879 5 g 5g
11378879 10 g 10g
11388879 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11358879 Supplier MP Biomedicals Supplier No. 0210204801
  • ONLINE EXCLUSIVE PRICE
    27.60 EUR
    26.25 EUR / 1g
    Save 1.35 EUR
    4% Off
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 830-96-6
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.21
MDL Number MFCD00005660
InChI Key GOLXRNDWAUTYKT-UHFFFAOYSA-N
Synonym 3-indolepropionic acid, indole-3-propionic acid, 3-1h-indol-3-yl propanoic acid, 1h-indole-3-propanoic acid, indolepropionic acid, indolylpropionic acid, 3-3-indolyl propionic acid, 3-3-indolyl propanoic acid, ipa auxin, 1h-indole-3-propionic acid
PubChem CID 3744
ChEBI CHEBI:43580
IUPAC Name 3-(1H-indol-3-yl)propanoic acid
SMILES OC(=O)CCC1=CNC2=CC=CC=C12

Specifications

Color Cream
Assay Percent Range ∽ 99%
Quantity 1 g
Formula Weight 189.21
Percent Purity ∼99%
Physical Form Powder
Chemical Name or Material Indole-3-propionic acid, Molecular biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.