Learn More
Indole-3-carbaldehyde, Sigma-Aldrich™
Specifications
Specifications
| CAS | 487-89-8 |
| Melting Point | 195°C to 198°C |
| MDL Number | MFCD00005622 |
| Quantity | 25 g |
| Synonym | Indole-3-carbaldehyde, Indol-3-aldehyde, Indole-3-carbaldehyde |
| Molecular Weight (g/mol) | 145.16 |
| Grade | Research |