missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-Butyric Acid Potassium Salt, MP Biomedicals™

Product Code. 11338879 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338879 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11338879 Supplier MP Biomedicals Supplier No. 0210204301

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Synonym: K-IBA, Potassium 4-(1H-Indol-3-yl)butanoate, 1H-Indole-3-Butanoic Acid Potassium Salt

Water-soluble form of IBA, an auxin-family plant growth hormone.

  • Promotes root organogenesis
  • Induces callus formation
  • Forms adventitious roots
  • Aids in regulation of gravitropism and phototropism
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C12H12KNO2
Molecular Weight (g/mol) 241.331
MDL Number MFCD00058442
InChI Key KTWDHJYSJOSTSJ-UHFFFAOYSA-M
Synonym potassium 4-1h-indol-3-yl butanoate, indole-3-butyric acid potassium salt, indole-3-butyric acid potassium, 4-indol-3-ylbutanoic acid, potassium salt, potassium indolebutyrate, pubchem22140, cambridge id 5119461, potassium ion indole-3 butyrate, indole-3-butyric acid-potassium salt, 3-indole butyric acid, potassium salt
PubChem CID 23664348
IUPAC Name potassium;4-(1H-indol-3-yl)butanoate
SMILES C1=CC=C2C(=C1)C(=CN2)CCCC(=O)[O-].[K+]

Specifications

CAS 60096-23-3
Quantity 1 g
Physical Form Powder

Unless specified otherwise, MP Biomedical™'s products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.