missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-butyric acid, MP Biomedicals™

Product Code. 11398869 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398869 1 g 1g
11308879 5 g 5g
11318879 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11398869 Supplier MP Biomedicals Supplier No. 0210204201
  • ONLINE EXCLUSIVE PRICE
    37.50 EUR
    35.70 EUR / 1g
    Save 1.80 EUR
    4% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 133-32-4
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.241
MDL Number MFCD00005664
InChI Key JTEDVYBZBROSJT-UHFFFAOYSA-N
Synonym indole-3-butyric acid, 3-indolebutyric acid, 4-1h-indol-3-yl butanoic acid, indolebutyric acid, hormodin, 1h-indole-3-butanoic acid, seradix, indole-3-butanoic acid, jiffy grow, 4-indol-3-yl butyric acid
PubChem CID 8617
ChEBI CHEBI:33070
IUPAC Name 4-(1H-indol-3-yl)butanoic acid
SMILES C1=CC=C2C(=C1)C(=CN2)CCCC(=O)O

Specifications

Color White
Quantity 1 g
Formula Weight 203.24
Physical Form Powder
Chemical Name or Material Indole-3-butyric acid, Plant growth hormone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.