Learn More
Indole-3-acetyl-L-aspartic Acid, TRC
Product Code. 30007304
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007304 25 mg 25mg
30004439 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30007304 Supplier Toronto Research Chemicals Supplier No. TRCI57737025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2456-73-7
Molecular Formula C14H14N2O5
Molecular Weight (g/mol) 290.27
Synonym N-[2-(1H-Indol-3-yl)acetyl]-L-aspartic Acid, N-(Indol-3-ylacetyl)-aspartic Acid, N-(Indol-3-ylacetyl)-L-aspartic Acid, (Indole-3-acetyl)aspartic Acid, (Indoleacetyl)aspartic Acid, 3-Indolylacetyl-L-aspartic Acid, Indol-3-ylacetylaspartic Acid, Indole-3-acetyl-L-aspartic Acid, Indole-3-acetylasparaginic Acid;
IUPAC Name (2S)-2-[[2-(1H-indol-3-yl)acetyl]amino]butanedioic acid
SMILES O=C(O)C[C@@H](C(O)=O)NC(CC1=CNC2=CC=CC=C21)=O
Specifications
InChI Formula InChI=1S/C14H14N2O5/c17-12(16-11(14(20)21)6-13(18)19)5-8-7-15-10-4-2-1-3-9(8)10/h1-4,7,11,15H,5-6H2,(H,16,17)(H,18,19)(H,20,21)/t11-/m0/s1
Quantity 25 mg
Formula Weight 290.0903
Shipping Condition Room Temperature
Chemical Name or Material Indole-3-acetyl-L-aspartic Acid