missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-acetic acid hydrazide, 98%, Thermo Scientific™

Product Code. 11437887
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11437887 5 g 5g
11427887 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11437887 Supplier Thermo Scientific Alfa Aesar Supplier No. A16562.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5448-47-5
Molecular Formula C10H11N3O
Molecular Weight (g/mol) 189.218
MDL Number MFCD00005634
InChI Key GYHLCXMCGCVVCG-UHFFFAOYSA-N
Synonym 2-1h-indol-3-yl acetohydrazide, indole-3-acetic acid hydrazide, 3-indoleacetic acid hydrazide, 1-3-indolylacetyl hydrazine, 3-indolylacetic acid hydrazide, 1h-indole-3-acetic acid, hydrazide, hydrazine, 1-3-indolylacetyl, indole 3-acethydrazide, indole-3-acetic hydrazide, indole-3-acetic acid, hydrazide
PubChem CID 21589
IUPAC Name 2-(1H-indol-3-yl)acetohydrazide
SMILES C1=CC=C2C(=C1)C(=CN2)CC(=O)NN

Specifications

Melting Point 142°C to 145°C
Quantity 5 g
Formula Weight 189.22
Percent Purity 98%
Chemical Name or Material Indole-3-acetic acid hydrazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.