accessibility menu, dialog, popup

UserName

CAS RN 5448-47-5

H2NNHOHN

CAS RN 5448-47-5

IUPAC Name: 2-(1H-indol-3-yl)acetohydrazide
Synonyms: 3-indoleacetic acid hydrazide
Molecular Weight (g/mol): 189.22
Molecular Formula: C10H11N3O
InChi Key: GYHLCXMCGCVVCG-UHFFFAOYSA-N
SMILES: NNC(=O)CC1=CNC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results