Learn More
Mettler Toledo™ Indicating Half-Cell Ion-Selective Electrodes
Product Code. 10312671
Change view
Click to view available options
pH Range:
2 to 9
4 to 10
Temperature (English) Operating:
0°F to 122°F
0°F to 176°F
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Temperature (English) Operating Unit Size
10312671 2 to 9 0°F to 122°F Each
10414641 4 to 10 0°F to 176°F Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10312671 Supplier Mettler Toledo™ Supplier No. 51340900
  • ONLINE EXCLUSIVE PRICE
    1300.00 EUR
    949.21 EUR / Pack of 1
    Save 350.79 EUR
    26% Off
View all promotions
In Stock
Requires
All ion-selective half cells require InLab™ 302 reference cell
For quick and easy determination of concentration of a variety of dissolved ions. Mettler Toledo Indicating Half-Cell Ion-Selective Electrodes have sturdy polyoxymethylene (POM) bodies.
  • Consist of a universal shaft and interchangeable ion-specific membrane module, allowing you to measure ion of your choice
  • All ion-selective half cells require InLab™ 302 reference cell (see 01-920-101, available separately), and appropriate S7 connection cable
Order Info
All are 4.75 in. long (120mm) and 0.5 in. (12mm) in diameter. For other ion-selective electrodes and membrane kits not listed here, contact your Fisher Scientific™ Customer Service Representative.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Determination of Ammonium concentration of samples such as boiler feed water, food and fertilizer
Material Polymer Matrix
Connector Types S7
Temperature (Metric) Operating 0°C to 50°C
Diameter (English) Body 0.5 in.
Diameter (Metric) Body 12 mm
Length (English) Body 4.75 in.
Length (Metric) Body 120 mm
pH Range 2 to 9
Temperature (English) Operating 0°F to 122°F
Concentration Measurement Range 1 to 1E-05mol/L