missing translation for 'onlineSavingsMsg'
Learn More

Inauhzin, MedChemExpress

Product Code. p-200097246
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496878 10 mg 10mg
16479417 5 mg 5mg
16476966 100 mg 100mg
16402947 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16496878 Supplier Medchem Express Supplier No. HY15869

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Inauhzin is a dual SirT1/IMPDH2 inhibitor, and acts as an activator p53, used in the research of cancer.

Inauhzin is a dual SirT1/IMPDH2 inhibitor, and acts as an activator p53, used in the research of cancer.

Specifications

Chemical Name or Material Inauhzin
Physical Form Solid
CAS 309271-94-1
Color Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C25H19N5OS2
For Use With (Application) Cancer-programmed cell death
Synonym INZ
Solubility Information DMSO : 21 mg/mL (44.72 mM; Need ultrasonic and warming)
SMILES CCC(SC1=NN=C2C(NC3=C2C=CC=C3)=N1)C(N4C5=C(C=CC=C5)SC6=CC=CC=C46)=O
Molecular Weight (g/mol) 469.58
Formula Weight 469.58
Percent Purity 98.05%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.