Learn More
Inauhzin, MedChemExpress
Product Code. 16496878
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16496878
Brand: Medchem Express HY15869
MedChemExpress Inauhzin is a dual SirT1/IMPDH2 inhibitor, and acts as an activator p53, used in the research of cancer.
Inauhzin is a dual SirT1/IMPDH2 inhibitor, and acts as an activator p53, used in the research of cancer.
Specifications
Chemical Name or Material Inauhzin
Physical Form Solid
CAS 309271-94-1
Color Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C25H19N5OS2
For Use With (Application) Cancer-programmed cell death
Synonym INZ
Solubility Information DMSO : 21 mg/mL (44.72 mM; Need ultrasonic and warming)
SMILES CCC(SC1=NN=C2C(NC3=C2C=CC=C3)=N1)C(N4C5=C(C=CC=C5)SC6=CC=CC=C46)=O
Molecular Weight (g/mol) 469.58
Formula Weight 469.58
Percent Purity 98.05%
Grade Research