missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Immuno Tubes and Stoppers

Product Code. 11371775
Change view
Click to view available options
Material:
Polyethylene
Polystyrene
Diameter (Metric) Outer:
11 mm
12 mm
15 mm
Product Type:
Immuno Star Tubes and Stopper
Immuno Tubes and Stopper
Stopper
For Use With (Equipment):
11 mm Nunc-Immuno Tubes
12 mm Nunc-Immuno Tubes
15 mm Nunc-Immuno Tubes
Immuno Tube
Unit Size:
Pack of 1200
Pack of 1800
Pack of 2400
Pack of 300
Pack of 3000
Pack of 3600
Pack of 500
Pack of 600
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Diameter (Metric) Outer Product Type For Use With (Equipment) Unit Size
11371775 Polyethylene 11 mm Stopper 11 mm Nunc-Immuno Tubes Pack of 600
10316141 Polyethylene 11 mm Stopper 11 mm Nunc-Immuno Tubes Pack of 3600
10172831 Polyethylene 15 mm Stopper 15 mm Nunc-Immuno Tubes Pack of 2400
11381775 Polyethylene 15 mm Stopper 15 mm Nunc-Immuno Tubes Pack of 300
11351785 Polyethylene 12 mm Stopper 12 mm Nunc-Immuno Tubes Pack of 500
10736662 Polyethylene 12 mm Stopper 12 mm Nunc-Immuno Tubes Pack of 3000
10254071 Polyethylene 12 mm Immuno Tubes and Stopper Immuno Tube Pack of 3000
10419962 Polystyrene 12 mm Immuno Tubes and Stopper Immuno Tube Pack of 3000
10596171 Polystyrene 11 mm Immuno Tubes and Stopper Immuno Tube Pack of 1800
10643452 Polyethylene 11 mm Immuno Tubes and Stopper - Pack of 3600
10419002 Polyethylene 15 mm Immuno Tubes and Stopper Immuno Tube Pack of 1200
10104421 Polystyrene 12 mm Immuno Star Tubes and Stopper Immuno Tube Pack of 3000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11371775 Leverantör Thermo Scientific™ Leverantörsnummer 341866

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

Reproduce results time after time with these immuno tubes and stoppers, ideal for ELISA, solid phase RIA and liquid phase RIA assays.

Reproduce results time after time with Thermo Scientific™ Immuno Tubes and Stoppers, ideal for ELISA, solid phase RIA and liquid phase RIA assays. These products are backed by industry-leading quality standards and certifications.

Features:
  • Offered in choice of three surfaces for optimal binding
  • Three tube sizes with stoppers available (ordered separately)
  • Fits standard equipment

Choice of Surface:

  • Thermo Scientific™ PolySorp™ is a hydrophobic surface that preferentially binds biomolecules that have hydrophobic domains.
  • Thermo Scientific™ MaxiSorp™ is a hydrophilic surface that is optimized for binding IgG and also displays increased binding of many other proteins and biomolecules that possess hydrophilic/hydrophobic character.
  • Thermo Scientific™ Mini-Sorp™ is a polyethylene-based material that provides a low-binding surface.

Liquid Phase Immunoassay Tubes (Cat. Nos. 443990, 468608):

  • MiniSorp surface
  • Molded from special polyethylene formulation
  • For liquid phase immunoasssays, including RIA, for storing of reagents, or collecting chromatographic fractions

Solid Phase Immunoassay Tubes:

  • PolySorp and MaxiSorp surfaces available for choice of optimal binding
  • For solid phase immuno techniques such as IRMA, ELISA and ILMA
  • Certified binding homogeneity
  • High–quality polystyrene for optimal quality
  • StarTube design gives higher sensitivity and faster assay times
  • Enhances assay sensitivity and reduces incubation times

Recommended for:

  • ELISA
  • Solid Phase RIA
  • Liquid Phase RIA
  • Sample storage
TRUSTED_SUSTAINABILITY

Specifikationer

Material Polyethylene
Product Type Stopper
Diameter (Metric) Outer 11 mm
No. per Pack 600
No. per Case 3600
Volume (Metric) Not Applicable
For Use With (Equipment) 11 mm Nunc-Immuno Tubes
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.